Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WBP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543795
Description
Peptide sequence: MADYWKSQPK KFCDYCKCWI ADNRPSVEFH ERGKNHKENV AKRISEIKQK Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
| WBP4 | |
| Polyclonal | |
| Unconjugated | |
| WBP4 | |
| AW545037; BB101031; domain-containing binding protein 4; FBP21; Fnbp21; formin binding protein 21; formin-binding protein 21; hypothetical protein LOC553723; MGC117310; Wbp4; WBP-4; WW domain binding protein 4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-binding protein 4-like protein; WW domain-containing binding protein 4; WW domain-containing-binding protein 4; zgc:112384; zgc:112384 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11193 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75554 | |
| WBP4 | |
| Synthetic peptide directed towards the N-terminal of human WBP4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction