Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558612 Supplier Invitrogen™ Supplier No. PA558612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YCKCWIADNR PSVEFHERGK NHKENVAKRI SEIKQKSLDK AKEEEKASKE FAAMEAAALK AYQEDLKRLG LESEILEPSI TPVTSTIPPT STSNQQKEK Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WBP4
Gene Accession No. O75554
Gene Alias AW545037; BB101031; domain-containing binding protein 4; FBP21; Fnbp21; formin binding protein 21; formin-binding protein 21; hypothetical protein LOC553723; MGC117310; Wbp4; WBP-4; WW domain binding protein 4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-binding protein 4-like protein; WW domain-containing binding protein 4; WW domain-containing-binding protein 4; zgc:112384; zgc:112384 protein
Gene Symbols WBP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBP4. Recombinant protein control fragment (Product #RP-97930).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.