Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WBP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566198
Description
Immunogen sequence: SDSDGEQEAEE GGVSTETEKP KIKFKEKNKN SDGGSDPETQ KEKSIQKQNS LGSNEEKSKT LKKSNPYGEW QEIKQ Highest antigen sequence identity to the following orthologs - mouse 66%, rat 62%.
This gene encodes WW domain-containing binding protein 4. The WW domain represents a small and compact globular structure that interacts with proline-rich ligands. This encoded protein is a general spliceosomal protein that may play a role in cross-intron bridging of U1 and U2 snRNPs in the spliceosomal complex A.
Specifications
WBP4 | |
Polyclonal | |
Unconjugated | |
WBP4 | |
AW545037; BB101031; domain-containing binding protein 4; FBP21; Fnbp21; formin binding protein 21; formin-binding protein 21; hypothetical protein LOC553723; MGC117310; Wbp4; WBP-4; WW domain binding protein 4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-binding protein 4-like protein; WW domain-containing binding protein 4; WW domain-containing-binding protein 4; zgc:112384; zgc:112384 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11193 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75554 | |
WBP4 | |
Recombinant Human WBP4. Recombinant protein control fragment (Product #RP-106884). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction