Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBSCR14 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3516R

Description
ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.Specifications
WBSCR14 | |
Polyclonal | |
Unconjugated | |
MLXIPL | |
BHLHD14; carbohydrate response element binding protein; Carbohydrate-responsive element-binding protein; ChREBP; Class D basic helix-loop-helix protein 14; Mio; MLX interacting protein like; MLX interacting protein-like; MLX interacting protein-like beta; MLX interactor; MLX-interacting protein-like; Mlxipl; MONDOB; putative hepatic transcription factor; Wbscr14; Williams Beuren syndrome chromosome region 14; Williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosomal region 14 protein homolog; Williams-Beuren syndrome chromosome region 14 homolog; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH | |
Rabbit | |
Protein A | |
RUO | |
51085 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NP71 | |
MLXIPL | |
KLH conjugated synthetic peptide derived from human WBSCR14/ChREBP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction