Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WBSCR16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563885 Supplier Thermo Scientific Supplier No. PA563885
Add to Cart
Add to Cart

WBSCR16 Polyclonal Antibody

WBSCR16 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rcc1l
Gene Accession No. Q96I51
Gene Alias 5730496C04Rik; AU019812; I79_011627; RCC1 like; RCC1L; RCC1-like; RCC1-like G exchanging factor-like protein; Wbscr16; Williams-Beuren syndrome chromosomal region 16 protein; Williams-Beuren syndrome chromosomal region 16 protein homolog; Williams-Beuren syndrome chromosomal region 16 protein-like; Williams-Beuren syndrome chromosome region 16; Williams-Beuren syndrome chromosome region 16 homolog; Williams-Beuren syndrome chromosome region 16 homolog (human)
Gene Symbols Rcc1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBSCR16.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.