Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBSCR22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131454
Description
WBSCR22 Polyclonal specifically detects WBSCR22 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
WBSCR22 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
WBSCR22 | |
Recombinant protein encompassing a sequence within the center region of human WBSCR22. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
114049 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.42mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
EC 2.1.1.-, FLJ44236, HASJ4442, HUSSY-3, MGC19709, MGC2022, MGC5140, PP3381, WBMT, Williams Beuren syndrome chromosome region 22, Williams Beuren syndrome chromosome region 22 protein, Williams-Beuren candidate region putative methyltransferase, Williams-Beuren syndrome chromosomal region 22 protein | |
Rabbit | |
32 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction