Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBSCR27 Polyclonal Antibody

Catalog No. PIPA554587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554587 Supplier Invitrogen™ Supplier No. PA554587

Rabbit Polyclonal Antibody

Immunogen sequence: LTTRTNSSNL QYKEALEATL DRLEQAGMWE GLVAWPVDRL WTAGSWLPPS WRWYPASLPR MASSPALSTC TESG Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 41%.

This gene encodes a protein belonging to ubiE/COQ5 methyltransferase family. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.22-q11.23. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR27
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL27
Gene Accession No. Q8N6F8
Gene Alias AW492986; BC002286; Methyltransferase-like protein 27; METTL27; WBSCR27; Williams Beuren syndrome chromosome region 27; Williams Beuren syndrome chromosome region 27 (human); Williams-Beuren syndrome chromosomal region 27 protein; Williams-Beuren syndrome chromosome region 27 homolog
Gene Symbols METTL27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBSCR27.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155368
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.