Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBSCR27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595711 Supplier Invitrogen™ Supplier No. PA595711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQEEGGSLP EVRARVRAAH GIPDLAQKLH FYDRWAPDYD QDVATLLYRA PRLAVDCLTQ ALPGPPHSAL ILDVACGTGL VAAELRAPGF LQLHGVDGSP GMLEQAQAPG LYQRLSLCTL GQEPLPSPEG TFDAVLIVGA LSDGQVPCNA IPELHVTKPG GLVCLTTRTN SSNLQYKEAL EATLDRLEQA GMWEGLVAWP VDRLWTAGSW LPPSWRWYPA SLPRMASSPA LSTCTESGRR PRLRK; Positive Samples: HT-29, MCF7, Mouse lung.

This gene encodes a protein belonging to ubiE/COQ5 methyltransferase family. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.22-q11.23. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR27
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene METTL27
Gene Accession No. Q8N6F8
Gene Alias AW492986; BC002286; Methyltransferase-like protein 27; METTL27; WBSCR27; Williams Beuren syndrome chromosome region 27; Williams Beuren syndrome chromosome region 27 (human); Williams-Beuren syndrome chromosomal region 27 protein; Williams-Beuren syndrome chromosome region 27 homolog
Gene Symbols METTL27
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-245 of human WBSCR27 (NP_6897722).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155368, 688407, 79565
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.