Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBSCR28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585023 Supplier Invitrogen™ Supplier No. PA585023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAAFEHTTQL AEAQEVEPQE VSGSSLLPSL SASSDSE Highest antigen sequence indentity to the following orthologs: Mouse - 51%, Rat - 50%.

WBSCR28 is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBSCR28
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM270
Gene Accession No. Q6UE05
Gene Alias MGC26719; TMEM270; Transmembrane protein 270; WBSCR28; Williams-Beuren syndrome chromosomal region 28 protein; Williams-Beuren syndrome chromosome region 28; Williams-Beuren syndrome critical region 28
Gene Symbols TMEM270
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TMEM270. Recombinant protein control fragment (Product #RP-108675).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.