Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WC1(+) gamma delta T cell Antibody (CACT21A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261083
Description
WC1(+) gamma delta T cell Monoclonal specifically detects WC1(+) gamma delta T cell in Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
WC1(+) gamma delta T cell | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
Mouse | |
Unpurified | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
CACT21A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
Bovine ConA stimulated mononuclear leukocytes | |
0.1 mg | |
Bovine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction