Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WC1 Monoclonal Antibody (19.19), FITC
GREENER_CHOICE

Catalog No. PIMA528517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528517 Supplier Invitrogen™ Supplier No. MA528517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WC1 Monoclonal Antibody for Flow

WC1 is approximately 215/300 kDa antigen expressed on the majority of gamma/delta T lymphocytes. These cells also express low levels of CD5, but are negative for other B and T cell markers. The WC1 antigen is reported to be involved in a number of processes, including activation of gamma/delta T cells and the development of a Th1-biased acquired immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WC1
Applications Flow Cytometry
Classification Monoclonal
Clone 19.19
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene LOC101108897
Gene Accession No. 0
Gene Alias antigen WC1.1; BoWC1.1; CD163 molecule-like 1; CD163L1; scavenger receptor cysteine-rich type 1 protein M160; scavenger-receptor protein; WC1; WC1 tail; WC1.1; WC1.1 molecule; WC1.1 tail; WC1-2; WC1-5; WC1-5 transmembrane protein
Gene Symbols LOC101108897
Host Species Mouse
Immunogen Ovine thymocytes.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101108897
Target Species Sheep
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.