Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDCP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178737
Description
WDCP Polyclonal specifically detects WDCP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
C2orf44 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
chromosome 2 open reading frame 44, FLJ21945, PP384 | |
Rabbit | |
Affinity Purified | |
RUO | |
80304 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
NP_079479.1 | |
WDCP | |
The immunogen for this product maps to a region between residue 450 and 500 of human Chromosome 2 Open Reading Frame 44 using the numbering given in entry NP_079479.1 (GeneID 80304). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction