Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDFY2 Polyclonal Antibody

Catalog No. PIPA588014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588014 Supplier Thermo Scientific Supplier No. PA588014

WDFY2 Polyclonal Antibody

WDFY2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen WDFY2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WDFY2
Gene Accession No. Q8BUB4, Q96P53
Gene Alias 5830485M08; B130024L21Rik; ProF; propeller-FYVE protein; WD repeat and FYVE domain containing 2; WD repeat and FYVE domain-containing protein 2; WD40 and FYVE domain containing 2; WD40- and FYVE domain-containing protein 2; WDF2; WDFY2; ZFYVE22; zinc finger FYVE domain-containing protein 22
Gene Symbols WDFY2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human WDFY2 (NP_4431821).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115825, 268752, 305956
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.