Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDFY3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP103332SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103332SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103332SS Supplier Novus Biologicals Supplier No. NBP103332SS

Rabbit Polyclonal Antibody has been used in 6 publications

WDFY3 Polyclonal specifically detects WDFY3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.

Specifications

Antigen WDFY3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunoprecipitation 2-5 mcg/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000, Electron Microscopy
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q8IZQ1
Gene Alias ALFY, Autophagy-linked FYVE protein, KIAA0993MGC16461, WD repeat and FYVE domain containing 3, WD repeat and FYVE domain-containing protein 3, ZFYVE25
Gene Symbols WDFY3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human WDFY3 protein (between amino acids 500-550) [Q8IZQ1]
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 23001.0
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.