Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDFY3 Monoclonal Antibody (4G9H4)
GREENER_CHOICE

Catalog No. FERMA548716
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548716 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548716 Supplier Invitrogen™ Supplier No. MA548716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WDFY3 Monoclonal Antibody for Western Blot, Flow, ELISA

The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDFY3
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4G9H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene WDFY3
Gene Accession No. Q8IZQ1
Gene Alias 2610509D04Rik; ALFY; Autophagy-linked FYVE protein; AW319683; B930017C24; Bchs; beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; D5Ertd66e; galactosyltransferase 3 beta 1, 4; Ggtb3; KIAA0993; LOW QUALITY PROTEIN: WD repeat and FYVE domain-containing protein 3; mKIAA0993; WD repeat and FYVE domain containing 3; WD repeat and FYVE domain-containing protein 3; WDFY3; ZFYVE25
Gene Symbols WDFY3
Host Species Mouse
Immunogen Purified recombinant fragment of human WDFY3 (AA: 3277-3526) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.