Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDFY3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569733
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 92%.
The ubiquitously expressed cellular upstream stimulatory factor (USF) consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.
Specifications
| WDFY3 | |
| Polyclonal | |
| Unconjugated | |
| WDFY3 | |
| 2610509D04Rik; ALFY; Autophagy-linked FYVE protein; AW319683; B930017C24; Bchs; beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; D5Ertd66e; galactosyltransferase 3 beta 1, 4; Ggtb3; KIAA0993; LOW QUALITY PROTEIN: WD repeat and FYVE domain-containing protein 3; mKIAA0993; WD repeat and FYVE domain containing 3; WD repeat and FYVE domain-containing protein 3; WDFY3; ZFYVE25 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23001 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IZQ1 | |
| WDFY3 | |
| synthetic peptide directed towards the C terminal region of human WDFY3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction