Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDFY4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559233 Supplier Invitrogen™ Supplier No. PA559233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMDNLKSQSP LPEQSPCLLP GFRVLNDFLA HHVHIPEVYL IVSTFFLQTP LTELMDGPKD SLDAMLQWLL QRHHQEEVLQ AGLCTEGALL LLEM Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.

WDFY4, also named as C10orf64 and KIAA1607, is a huge protein with unknown functions. WDFY4 is expressed predominantly in immune tissues such as lymph nodes, the spleen, the thymus and the tonsils. According to recent genome-wide association studies (GWAS), WDFY4 was introduced as novel predisposing genes for SLE (Systemic Lupus Erythematosus) which is a heterogeneous complex relapsing-remitting autoimmune disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDFY4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Wdfy4
Gene Accession No. Q6ZS81
Gene Alias C10orf64; Gm18810; KIAA1607; RGD1564142; WD repeat and FYVE domain containing 4; WD repeat- and FYVE domain-containing protein 4; WDFY family member 4; Wdfy4
Gene Symbols Wdfy4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDFY4. Recombinant protein control fragment (Product #RP-97077).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57705
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.