Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540609 Supplier Invitrogen™ Supplier No. PA540609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KQILHGDPLP LTRKSYLAWI GFSAEGTPCY VDSEGIVRML NRGLGNTWTP Sequence homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.

The protein encoded by this gene contains multiple N-terminal WD40 domains and a C-terminal high mobility group (HMG) box. WD40 domains are found in a variety of eukaryotic proteins and may function as adaptor/regulatory modules in signal transduction, pre-mRNA processing and cytoskeleton assembly. HMG boxes are found in many eukaryotic proteins involved in chromatin assembly, transcription and replication. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDHD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Wdhd1
Gene Accession No. O75717
Gene Alias A630092E18; Acidic nucleoplasmic DNA-binding protein 1; AND1; And-1; CHTF4; crm1; CRM1/XPO1 protein; CTF4; CTF4, chromosome transmission fidelity factor 4 homolog; D630024B06Rik; exportin 1 (CRM1 homolog); exportin 1 S homeolog; exportin-1; WD repeat and HMG-box DNA binding protein 1; WD repeat and HMG-box DNA-binding protein 1; WD repeat and HMG-box DNA-binding protein 1-like protein; WDHD1; XELAEV_18028477mg; Xpo; xpo1; xpo1.S
Gene Symbols Wdhd1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human WDHD1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11169
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.