Learn More
Invitrogen™ WDHD1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total WDHD1.
Specifications
Specifications
| Antigen | WDHD1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Wdhd1 |
| Gene Accession No. | O75717 |
| Gene Alias | A630092E18; Acidic nucleoplasmic DNA-binding protein 1; AND1; And-1; CHTF4; crm1; CRM1/XPO1 protein; CTF4; CTF4, chromosome transmission fidelity factor 4 homolog; D630024B06Rik; exportin 1 (CRM1 homolog); exportin 1 S homeolog; exportin-1; WD repeat and HMG-box DNA binding protein 1; WD repeat and HMG-box DNA-binding protein 1; WD repeat and HMG-box DNA-binding protein 1-like protein; WDHD1; XELAEV_18028477mg; Xpo; xpo1; xpo1.S |
| Gene Symbols | Wdhd1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.