Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDPCP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560627 Supplier Invitrogen™ Supplier No. PA560627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIIFQYIHCD EIYEAINILS SMNWDTLGHQ CFISMSAIVN HLLRQKLTPE REAQLETSLG TFYAPTRPLL DSTILEYRDQ ISKYARRF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 80%.

The C2ORF86 gene encodes an ortholog of Drosophila 'fritz,' a cytoplasmic WD40 repeat protein that controls planar cell polarity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDPCP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDPCP
Gene Accession No. O95876
Gene Alias AV249152; Bardet-Biedl syndrome 15 protein; BBS15; C2orf86; CHDTHP; FRITZ; FRTZ; hFRTZ; homoloc-13; homolog-13; mFrtz; WD repeat containing planar cell polarity effector; WD repeat-containing and planar cell polarity effector protein; WD repeat-containing and planar cell polarity effector protein fritz homolog; WD repeat-containing protein C2orf86; WD repeat-containing protein C2orf86 homolog; Wdpcp
Gene Symbols WDPCP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDPCP. Recombinant protein control fragment (Product #RP-99655).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.