Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220887
Description
WDR1 Polyclonal specifically detects WDR1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
WDR1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Actin-interacting protein 1, AIP1, NORI-1, WD repeat domain 1, WD repeat-containing protein 1 | |
Rabbit | |
66 kDa | |
0.1 mL | |
Stem Cell Markers | |
9948 | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.89mg/mL | |
Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O75083 | |
WDR1 | |
Recombinant protein encompassing a sequence within the center region of human WDR1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat. Species reactivity based on sequence identity: Mouse (97%), Chicken (94%), Dog (99%), Zebrafish (82%), Bovine (97%), Xenopus tropicalis (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction