Learn More
Invitrogen™ WDR1 Polyclonal Antibody

Description
Recommended positive controls: H1299, rat brain. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (82%), Xenopus laevis (88%), Dog (99%), Chicken (94%), Chimpanzee (100%), Bovine (97%), Guinea pig (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | WDR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | WDR1 |
| Gene Accession No. | O75083, Q5RKI0 |
| Gene Alias | Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1 |
| Gene Symbols | WDR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.