Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557657 Supplier Invitrogen™ Supplier No. PA557657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EHLLKYEYQP FAGKIKDIAW TEDSKRIAVV GEGREKFGAV FLWDSGSSVG EITGHNKVIN SVDIKQSRPY RLATGSDDNC AAFFEGPPFK FKFTI Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR1
Gene Accession No. O75083
Gene Alias Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1
Gene Symbols WDR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR1. Recombinant protein control fragment (Product #RP-96602).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9948
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.