Learn More
Invitrogen™ WDR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588389
Description
Immunogen sequence: YINYLDRNNP SKPLHVIKGH SKSIQCLTVH KNGGKSYIYS GSHDGHINYW DSETGENDSF AGKGHTNQVS RMTVDESGQL ISCSMDDTVR YTSLMLRDYS GQGVVKLDVQ PKCVAVGPGG YAVVVCIGQI VLLKDQRKCF SIDNPGYEPE VVAVHPGGDT VAIGGVDGNV RLYSILGTTL KDEGKLLEAK GPVTDVAYSH D; Positive Samples: Raji, HT-29, U-87MG, MCF7, NIH/3T3, Mouse brain, Mouse liver, Rat heart, Rat spinal cord; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, podosome.
This gene encodes a protein containing 9 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, mostly including a trp-asp at the C-terminal end. WD domains are involved in protein-protein interactions. The encoded protein may help induce the disassembly of actin filaments. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
WDR1 | |
Polyclonal | |
Unconjugated | |
WDR1 | |
Actin-interacting protein 1; Aip1; D5Wsu185e; epididymis secretory protein Li 52; HEL-S-52; NORI-1; rede; WD repeat domain 1; WD repeat protein 1; WD repeat-containing protein 1; WD40 repeat protein 1; Wdr1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22388, 360950, 9948 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.3 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75083, O88342, Q5RKI0 | |
WDR1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 300-500 of human WDR1 (NP_0598301). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.