Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541985 Supplier Invitrogen™ Supplier No. PA541985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DTRSCKAPLY DLAAHEDKVL SVDWTDTGLL LSGGADNKLY SYRYSPTTSH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 89%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein is highly similar to the mouse WD repeat domain 12 protein at the amino acid level. The protein encoded by this gene is a component of a nucleolar protein complex that affects maturation of the large ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene WDR12
Gene Accession No. Q9GZL7
Gene Alias 4933402C23Rik; fb24f09; HAMAP-Rule:MF_03029}; MNCb-5414; nuclear protein Ytm1; ribosome biogenesis protein wdr12; ribosome biogenesis protein WDR12 {ECO:0000255; WD repeat domain 12; WD repeat domain 12 protein; WD repeat-containing protein 12; WD repeat-containing protein 12 {ECO:0000255; WDR12; wdr12 protein; wu:fb24f09; YTM1; Ytm1p; zgc:55609
Gene Symbols WDR12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human WDR12 (aa 371-420).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.