Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584203 Supplier Invitrogen™ Supplier No. PA584203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTPPLLCGKV SRDIRQEIEK LTANSQVKKL RWFSECLSPP GGSDNLWNLV AVIKGQDDSL LPQNYCKGIM HLKHLIKFRT SEAQELTTVK M Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.

This gene encodes a WD repeat-containing protein. It is abundantly expressed in retina and testis, and is thought to be a candidate gene for retinal disease. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Nov 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR17
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR17
Gene Accession No. Q8IZU2
Gene Alias WD repeat domain 17; WD repeat-containing protein 17; WDR17
Gene Symbols WDR17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR17. Recombinant protein control fragment (Product #RP-108404).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.