Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562153
Description
Immunogen sequence: LPHFNKHLLG AEHGDEPRHG GLTLRLGLHQ QGSEPSYLDR TEQLQAVLCS TMEKSVLGGQ DQLRVRVTEL EDEVRNLRKI NRDLFDFST Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
WD repeat domain 18 (WDR18) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WD repeats are minimally conserved regions of approximately 40 amino acids that commonly form a tertiary propeller structure. WDR18 is a 432 amino acid protein that contains six WD repeats and is encoded by a gene located on human chromosome 19. WDR18 may play a role during development. The complex PELP1-TEX10-WDR18 is required for the maturation and nucleolar release of the large ribosomal subunit.
Specifications
WDR18 | |
Polyclonal | |
Unconjugated | |
WDR18 | |
2310012I10Rik; AU044733; AW122032; Involved in Processing ITS2 3 homolog; Ipi3; R32184_1; WD repeat domain 18; WD repeat-containing protein 18; WDR18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57418 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.07 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV38 | |
WDR18 | |
Recombinant protein corresponding to Human WDR18. Recombinant protein control fragment (Product #RP-98891). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction