Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110578 Supplier Invitrogen™ Supplier No. PA5110578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLFTWPGQRE RSFHPEQDAG KVFKGHRNQV TCLSVSTDGS VLLSGSHDET VRLWDVQSKQ CIRTVALKGP VTNAAILLAP VSMLSSDFRP SLPLPHFNKH LLGAEHGDEP RHGGLTLRLG LHQQGSEPSY LDRTEQLQAV LCSTMEKSVL GGQDQLRVRV TELEDEVRNL RKINRDLFDF STRFITRPAK.

WD repeat domain 18 (WDR18) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WD repeats are minimally conserved regions of approximately 40 amino acids that commonly form a tertiary propeller structure. WDR18 is a 432 amino acid protein that contains six WD repeats and is encoded by a gene located on human chromosome 19. WDR18 may play a role during development. The complex PELP1-TEX10-WDR18 is required for the maturation and nucleolar release of the large ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR18
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene WDR18
Gene Accession No. Q499N3, Q9BV38
Gene Alias 2310012I10Rik; AU044733; AW122032; Involved in Processing ITS2 3 homolog; Ipi3; R32184_1; WD repeat domain 18; WD repeat-containing protein 18; WDR18
Gene Symbols WDR18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 243-432 of human WDR18 (NP_077005.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314617, 57418
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.