Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531388 Supplier Invitrogen™ Supplier No. PA531388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (80%), Rat (80%), Rhesus Monkey (95%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains six WD repeats, a clathrin heavy-chain repeat, and three transmembrane domains. This gene is conserved from C. elegans to human. It may participate in androgen-regulated signaling mechanisms or in the vesicular trafficking of androgen-regulated secretory processes. Alternatively spliced transcript variants encoding distinct isoforms have been reported but the full-length nature of one of these variants has not been defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene WDR19
Gene Accession No. Q3UGF1, Q8NEZ3
Gene Alias ATD5; C330027H04Rik; CED4; D330023L08Rik; DYF2; DYF-2; Ift144; Intraflagellar transport 144 homolog; Kiaa1638; mKIAA1638; NPHP13; ORF26; Oseg6; PWDMP; RGD1306997; SRTD5; WD repeat domain 19; WD repeat membrane protein PWDMP; WD repeat-containing protein 19; WDR19
Gene Symbols WDR19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human WDR19. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213081, 57728
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.