Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR19 Polyclonal Antibody, Abbomax

Catalog No. 502204084
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4084 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4084 Supplier Abbomax Inc Supplier No. 602550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Testis tissue lysate Cellular location: Cytoplasm, cytoskeleton, cilium basal body.

WDR19 Polyclonal antibody specifically detects WDR19 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen WDR19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene WDR19
Gene Accession No. Q3UGF1, Q8NEZ3
Gene Alias ATD5; C330027H04Rik; CED4; D330023L08Rik; DYF2; DYF-2; Ift144; Intraflagellar transport 144 homolog; Kiaa1638; mKIAA1638; NPHP13; ORF26; Oseg6; PWDMP; RGD1306997; SRTD5; WD repeat domain 19; WD repeat membrane protein PWDMP; WD repeat-containing protein 19; WDR19
Host Species Rabbit
Immunogen A synthetic peptide 880-920 amino acids of WDR19 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213081, 305349, 57728
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.