Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548912
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548912 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548912 Supplier Invitrogen™ Supplier No. PA548912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR20 Polyclonal Antibody for Western Blot

CUL-2 is a member of the family of human cullin genes (CUL-1, -2, -3, -4a, -4b, and -5), homologous to the S. cerevisiae cdc53 gene. CUL-1 forms a complex with human p19skp1 and F box protein p45skp2. Together with cdc53 (Known as CulA), as three subunits, they form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL-1, CUL-2 does not associate with p19skp1 and p45skp2. Instead, CUL-2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR20
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene WDR20
Gene Accession No. Q8TBZ3
Gene Alias 2310040A13Rik; AI549910; DMR; Protein DMR; WD repeat domain 20; WD repeat domain 20A; WD repeat-containing protein 20; WDR20; Wdr20a
Gene Symbols WDR20
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 5-32 amino acids from the N-terminal region of human WDR20.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.