Learn More
WDR21A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301907AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The nuclear protein p100 is a proliferation-associated protein whose expression is restricted to cell cycle phases S, G2, and M. Exclusively expressed in proliferating cells from the transition G1/S until the end of cytokinesis. During mitosis it is strictly associated with the spindle pole and with the mitotic spindle, whereas during S and G2, it is diffusely distributed throughout the nucleus. The full-length cDNA encodes a 747-amino acid protein with a putative ATP/GTP-binding site motif. RT-PCR analysis demonstrated strong expression of in lung carcinoma cell lines but not in normal lung tissues. Expression was also found in adult placenta, skeletal muscle, thymus, testis, and small intestine and in fetal brain, liver, and kidney. P100 is also correlated to cancer prognosis.Specifications
WDR21A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8WV16 | |
Dcaf4 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DCAF4 | |
DKFZp434K114, WD repeat domain 21, WD repeat domain 21A, WD repeat-containing protein 21A, WDR21, WDR21A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26094, 73828 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.