Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561650
Description
Immunogen sequence: LWHMEQAIHS RVKPIQLKGE HHSNIFCLAF NSGNTKVFSG GNDEQVILHD VESSETLDVF AHEDAVYGLS VSPVNDNIFA SSSDDG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
WDR22 | |
Polyclonal | |
Unconjugated | |
DCAF5 | |
BCRG2; BCRP2; Breakpoint cluster region protein 2; breakpoint cluster region protein, uterine leiomyoma, 2; D14S1461E; DCAF5; DDB1 and CUL4 associated factor 5; DDB1- and CUL4-associated factor 5; KIAA1824; WD repeat-containing protein 22; WDR22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JK2 | |
DCAF5 | |
Recombinant protein corresponding to Human WDR22. Recombinant protein control fragment (Product #RP-98471). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction