Learn More
WDR23 Polyclonal Antibody

Description
WDR23 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
Specifications
| Antigen | WDR23 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Dcaf11 |
| Gene Accession No. | Q8TEB1 |
| Gene Alias | 0710008A13Rik; C76035; D14Ucla1; Dacf11; DCAF11; DDB1 and CUL4 associated factor 11; DDB1- and CUL4-associated factor 11; DKFZp779A1629; GL014; GLO14; PRO2389; WD repeat domain 23; WD repeat domain 23 isoform 1; WD repeat-containing protein 23; WDR23; WDR23 protein |
| Gene Symbols | Dcaf11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.