Learn More
Invitrogen™ WDR23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590526
Description
Immunogen sequence: MGSRNSSSAG SGSGDPSEGL PRRGAGLRRS EEEEEEDEDV DLAQVLAYLL RRGQVRLVQG GGAANLQFIQ ALLDSEEEND RAWDGRLGDR YNPPVDATPD TRELEFNEIK TQVELATGQL GLRRAAQKHS FPRMLHQRER GLCHRGSFSL GEQSRVISHF LPNDLGFTDS; Positive Samples: LO2, HeLa, Mouse brain, Mouse heart, Rat brain.
WDR23 contains 7 WD repeats. The function of WDR23 remains unknown.This gene encodes a WD repeat-containing protein. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.
Specifications
WDR23 | |
Polyclonal | |
Unconjugated | |
Dcaf11 | |
0710008A13Rik; C76035; D14Ucla1; Dacf11; DCAF11; DDB1 and CUL4 associated factor 11; DDB1- and CUL4-associated factor 11; DKFZp779A1629; GL014; GLO14; PRO2389; WD repeat domain 23; WD repeat domain 23 isoform 1; WD repeat-containing protein 23; WDR23; WDR23 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
28199, 305895, 80344 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.63 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q5M9G8, Q8TEB1, Q91VU6 | |
Dcaf11 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human DCAF11 (NP_0795063). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.