Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR23 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988520
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8520 Supplier Bioss Supplier No. BS8388R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR23 contains 7 WD repeats. The function of WDR23 remains unknown.This gene encodes a WD repeat-containing protein. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.

Specifications

Antigen WDR23
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Dcaf11
Gene Accession No. Q8TEB1
Gene Alias 0710008A13Rik; BOS_10208; C76035; D14Ucla1; Dacf11; Dcaf11; DDB1 and CUL4 associated factor 11; DDB1- and CUL4-associated factor 11; DKFZP459A2062 protein-like; DKFZp779A1629; GL014; GLO14; LOC100342985; PRO2389; WD repeat domain 23; WD repeat-containing protein 23; Wdr23
Gene Symbols Dcaf11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human WDR23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80344
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.