Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR26 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126630H
Description
WDR26 Polyclonal antibody specifically detects WDR26 in Human samples. It is validated for Western BlotSpecifications
WDR26 | |
Polyclonal | |
Western Blot | |
MIP2, myocardial ischemic preconditioning up-regulated protein 2, WD repeat domain 26, WD repeat-containing protein 26 | |
The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human WD repeat domain 26 using the numbering given in entry BAG10986.1 (GeneID 80232). | |
0.1 mL | |
Cell Biology, Endocrinology, Signal Transduction | |
80232 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction