Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR26 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562121
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2121 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2121 Supplier Bethyl Laboratories, Inc Supplier No. A302244AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and X. tropicalis

May be involved in MAPK pathways.

Specifications

Antigen WDR26
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene WDR26
Gene Accession No. Q9H7D7
Gene Alias CDW2, CUL4- and DDB1-associated WDR protein 2, FLJ21016, GID complex subunit 7 homolog, GID complex subunit 7 homolog (S. cerevisiae), GID7, MIP2, myocardial ischemic preconditioning upregulated protein 2, Myocardial ischemic preconditioning up-regulated protein 2
Gene Symbols WDR26
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80232
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.