Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077296
Description
WDR3 Polyclonal specifically detects WDR3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
WDR3 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
dJ776P7.2 (WD repeat domain 3), FLJ12796, WD repeat domain 3, WD repeat-containing protein 3 | |
Rabbit | |
106 kDa | |
0.1 mL | |
Apoptosis | |
10885 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
Q9UNX4 | |
WDR3 | |
The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human WD repeat-containing domain 3 using the numbering given in entry NP_006775.1 (GeneID 10885). | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction