Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554998 Supplier Invitrogen™ Supplier No. PA554998
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GARLFGWLKE RSLGRGLFVD PARDNFRTMT SLYGSIHPAD SVYLSTRTHG AVFNLEYSPD GSVLTVACEQ TEVLL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

DCAF10 may function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR32
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DCAF10
Gene Accession No. Q5QP82
Gene Alias AA959934; DCAF10; DDB1 and CUL4 associated factor 10; DDB1- and CUL4-associated factor 10; RGD1304730; WD repeat domain 32; WD repeat-containing protein 32; WDR32
Gene Symbols DCAF10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR32. Recombinant protein control fragment (Product #RP-92941).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79269
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.