Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR33 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565445 Supplier Invitrogen™ Supplier No. PA565445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAWHPLGHILC SGSNDHTSKF WTRNRPGDKM RDRYNLNLLP GMSEDGVEYD DLEPNSLAVI PGMGIPEQLK LAMEQEQMGK DESNEIEMTI PGLDWGMEEV MQKDQKKVPQ KKVPYAKPIP AQFQQAWMQN KVPIP Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

WDR33 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is highly expressed in testis and the protein is localized to the nucleus. This gene may play important roles in the mechanisms of cytodifferentiation and/or DNA recombination. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR33
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Wdr33
Gene Accession No. Q9C0J8
Gene Alias 1110001N06Rik; 2310011G05Rik; 2810021O11Rik; 8430413N20Rik; FLJ11294; FLJ11294-like; NET14; pre-mRNA 3' end processing protein WDR33; WD repeat domain 33; WD repeat-containing protein 33; WD repeat-containing protein of 146 kDa; WD repeat-containing protein WDC146; WDC146; Wdr33; WD-repeat protein 33; zgc:110570
Gene Symbols Wdr33
Host Species Rabbit
Immunogen Recombinant Human WDR33. Recombinant protein control fragment (Product #RP-106125).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55339
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.