Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR34 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | WDR34 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
WDR34 Polyclonal specifically detects WDR34 in Human samples. It is validated for Western Blot.Specifications
WDR34 | |
Polyclonal | |
Rabbit | |
Q96EX3 | |
89891 | |
Synthetic peptides corresponding to WDR34(WD repeat domain 34) The peptide sequence was selected from the C terminal of WDR34. Peptide sequence SLKYLFAVRWSPVRPLVFAAASGKGDVQLFDLQKSSQKPTVLIKQTQDES. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
bA216B9.3, MGC20486, WD repeat domain 34, WD repeat-containing protein 34 | |
WDR34 | |
IgG | |
58 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title