Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116177
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116177 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116177 Supplier Invitrogen™ Supplier No. PA5116177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

WDR34 Polyclonal Antibody for Western Blot

WDR34 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation.This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR34
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene WDR34
Gene Accession No. Q96EX3
Gene Alias 3200002I06Rik; bA216B9.3; DIC5; DYNC2I2; dynein 2 intermediate chain 2; FAP133; MGC20486; RP11-216B9.5; SRTD11; WD repeat domain 34; WD repeat-containing protein 34; WDR34
Gene Symbols WDR34
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 305-334 amino acids from the Central region of human WDR34.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89891
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.