Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR35 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282058
Description
WDR35 Polyclonal specifically detects WDR35 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
WDR35 | |
Polyclonal | |
Unconjugated | |
PBS | |
WDR35 | |
WDR35 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human WDR35. The immunogen is located within amino acids 60 - 110 of WDR35. Amino Acid Squence: TWNEQYQKLTTSDENG | |
0.1 mg | |
Primary | |
WDR35 antibody is human specific. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin | |
CED2, IFT121, WD repeat domain 35 | |
Rabbit | |
Affinity Purified | |
RUO | |
57539 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction