Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDR35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560629
Description
Immunogen sequence: SLPNVGLIQK YSLNCRAYQL SLNCNSSRLA IIDISGVLTF FDLDARVTDS TGQQVVGELL KLERRDVWDM KWAKDNPDLF AMMEKTRMYV FRNLDPEEPI QTSGY Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR35, also known as CED2, has been shown to be mutated in patients with Sensenbrenner syndrome/cranioectodermal dysplasia (CED), an autosomal-recessive disease that is characterized by craniosynstosis and ectodermal and skeletal abnormalities. WDR35 localizes to cilia and dentrosomes during embryogenesis and human and mouse fibroblasts that lack this gene fail to produce cilia. Mutations in this gene can also cause short-rib polydactyly syndromes due to abnormal ciliogenesis.
Specifications
| WDR35 | |
| Polyclonal | |
| Unconjugated | |
| WDR35 | |
| 4930459M12Rik; 4931430C06; CED2; IFT121; IFTA1; intraflagellar transport protein 121 homolog; KIAA1336; mKIAA1336; Naofen; RGD1564116; SRTD7; WD repeat domain 35; WD repeat domain 35-like; WD repeat-containing protein 35; WDR35; Wdr35l | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57539 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2L0 | |
| WDR35 | |
| Recombinant protein corresponding to Human WDR35. Recombinant protein control fragment (Product #RP-99642). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction