Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534370
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534370 Supplier Invitrogen™ Supplier No. PA534370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human testis tissue. PA5-3437°Can be used with blocking peptide PEP-1413.

WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR35, also known as CED2, has been shown to be mutated in patients with Sensenbrenner syndrome/cranioectodermal dysplasia (CED), an autosomal-recessive disease that is characterized by craniosynstosis and ectodermal and skeletal abnormalities. WDR35 localizes to cilia and dentrosomes during embryogenesis and human and mouse fibroblasts that lack this gene fail to produce cilia. Mutations in this gene can also cause short-rib polydactyly syndromes due to abnormal ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR35
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene WDR35
Gene Accession No. Q9P2L0
Gene Alias 4930459M12Rik; 4931430C06; CED2; IFT121; IFTA1; intraflagellar transport protein 121 homolog; KIAA1336; mKIAA1336; Naofen; RGD1564116; SRTD7; WD repeat domain 35; WD repeat domain 35-like; WD repeat-containing protein 35; WDR35; Wdr35l
Gene Symbols WDR35
Host Species Rabbit
Immunogen A 16 amino acid synthetic peptide near the amino terminus of human WDR35.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57539
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.