Learn More
Invitrogen™ WDR36 Polyclonal Antibody

Description
The antibody detects endogenous levels of total WDR36 protein.
Specifications
Specifications
| Antigen | WDR36 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WDR36 |
| Gene Accession No. | Q8NI36 |
| Gene Alias | GLC1G; TAWDRP; TA-WDRP; T-cell activation WD repeat protein; T-cell activation WD repeat-containing protein; UTP21; WD repeat domain 36; WD repeat-containing protein 36; WDR36 |
| Gene Symbols | WDR36 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.