Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558111 Supplier Invitrogen™ Supplier No. PA558111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKSLGHGLIN KKRVKRKGLQ NTMSVRLPPI TKFAAEEARE SDWDGIIACH QGKLSCSTWN YQKSTIGAYF LKPKELKKDD ITATAVDITS CGNF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Mutations in this gene have been associated with adult-onset primary open-angle glaucoma (POAG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR36
Gene Accession No. Q8NI36
Gene Alias GLC1G; TAWDRP; TA-WDRP; T-cell activation WD repeat protein; T-cell activation WD repeat-containing protein; UTP21; WD repeat domain 36; WD repeat-containing protein 36; WDR36
Gene Symbols WDR36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR36. Recombinant protein control fragment (Product #RP-96879).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.