Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596650 Supplier Invitrogen™ Supplier No. PA596650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKDSLVLLGR VPAHPDSRCW FLAWNPAGTL LASCGGDRRI RIWGTEGDSW ICKSVLSEGH QRTVRKVAWS PCGNYLASAS FDATTCIWKK NQDDFECVTT LEGHENEVKS VAWAPSGNLL ATCSRDKSVW VWEVDEEDEY ECVSVLNSHT QDVKHVVWHP SQELLASASY DDTVKLYREE EDDWVCCATL EGHESTVWSL AFDPSGQRLA SCSDDRTVRI WRQYLPGNEQ GVACSGSDPS WKCICTLSGF HSRTIYDIAW CQLTGALATA CGDDAIRVFQ EDPNSDPQQP TFSLTAHLHQ AHSQDVNCVA WNPKEPGLLA SCSDDGEVAF WKYQRPEGL; Positive Samples: Jurkat, Mouse kidney, Mouse thymus, Mouse heart, Rat heart.

WDR39 is a member of the WD40 family of proteins. WDR39 specifically interacts with WT1 both in vitro and in vivo. This interaction results in a decrease in transcriptional activation mediated by WT1. WDR39 does not inhibit binding of WT1 to its consensus nucleotide sequence and does not affect the repression activity of WT1. Thus, WDR39 appears to specifically modulate the transactivation activity of WT1 and may function to regulate the physiological functions of WT1 in cell growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR39
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CIAO1
Gene Accession No. O76071, Q5M7T1, Q99KN2
Gene Alias AW210570; CIA1; ciao1; ciao1 {ECO:0000255; cytosolic iron-sulfur assembly component 1; cytosolic iron-sulfur protein assembly 1; cytosolic iron-sulfur protein assembly 1 homolog; HAMAP-Rule:MF_03037}; probable cytosolic iron-sulfur protein assembly protein CIAO1; probable cytosolic iron-sulfur protein assembly protein CIAO1 {ECO:0000255; WD repeat domain 39; WD repeat-containing protein 39; WD repeat-containing protein 39 {ECO:0000255; WD40 protein Ciao1; Wdr39
Gene Symbols CIAO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-339 of human CIAO1 (NP_0047951).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26371, 29231, 9391
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.