Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR4 Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
SDP

Catalog No. 502540693
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0693 30 μL
50-167-4614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0693 Supplier OriGene Supplier No. TA503815S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WDR4 Monoclonal antibody specifically detects WDR4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen WDR4
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4G1
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Katnb1
Gene Accession No. Q9BVA0
Gene Alias AI415180; AI448349; D530049K22Rik; hWH; mWH; Protein Wuho homolog; TRM82; TRM82 tRNA methyltransferase 82 homolog; TRMT82; tRNA (guanine-N(7)-)-methyltransferase non-catalytic subunit WDR4; tRNA (guanine-N(7)-)-methyltransferase subunit WDR4; WD repeat domain 4; WD repeat-containing protein 4; WD40 repeat protein 4; WDR4
Host Species Mouse
Immunogen Full length human recombinant protein of human KATNB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10300
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.